Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 96%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
69304-37-6
C12H28Cl2OSi2
315.425
MFCD00009655
DDYAZDRFUVZBMM-UHFFFAOYSA-N
1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane,1,3-dichlorotetraisopropyldisiloxane,1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane,1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane,disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl,1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane,chloro chlorodiisopropylsilyl oxy diisopropylsilane,chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane,tipdsicl2,tipdsi-cl2
172404
chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane
CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl
69304-37-6
C12H28Cl2OSi2
315.425
MFCD00009655
DDYAZDRFUVZBMM-UHFFFAOYSA-N
1,3-dichloro-1,1,3,3-tetraisopropyldisiloxane,1,3-dichlorotetraisopropyldisiloxane,1,1,3,3-tetraisopropyl-1,3-dichlorodisiloxane,1,3-dichloro-1,1,3,3-tetraisopropyl-disiloxane,disiloxane, 1,3-dichloro-1,1,3,3-tetrakis 1-methylethyl,1,1,3,3-tetraisopropyl-1,3-dichlorosiloxane,chloro chlorodiisopropylsilyl oxy diisopropylsilane,chloro-chloro-di propan-2-yl silyl oxy-di propan-2-yl silane,tipdsicl2,tipdsi-cl2
172404
chloro-[chloro-di(propan-2-yl)silyl]oxy-di(propan-2-yl)silane
CC(C)[Si](C(C)C)(O[Si](C(C)C)(C(C)C)Cl)Cl
Refractive Index 1.4533 to 1.4553
Color Colorless
Flash Point 76°C
Packaging Glass bottle
Boiling Point 120.0°C (15.0 mmHg)
Infrared Spectrum Authentic
Density 1.0000g/mL
Quantity 5mL
CAS Min % 95.0
CAS Max % 100.0
Assay Percent Range 96%
Linear Formula [(CH3)2CH]2Si(Cl)OSi(Cl)[CH(CH3)2]2
Fieser 16,125; 17,121
Formula Weight 315.42
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material 1,3-Dichloro-1,1,3,3-tetraisopropyldisiloxane, 96%
Show More Show Less

GHS P Statement
Causes severe skin burns and eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

TSCA : TSCA