Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dichloro-2-fluorobenzene, 98%

Click to view available options
Quantity:
1g
5g
25g
2274-11-5
C8H10O4
170.16
MFCD00010307
KUDUQBURMYMBIJ-UHFFFAOYSA-N
ethylene diacrylate,ethylene acrylate,ethylene glycol diacrylate,acrylic acid, ethylene ester,acrylic acid, ethylene glycol diester,2-propenoic acid, 1,2-ethanediyl ester,acrylic acid, diester with ethylene glycol,1,2-ethanediyl diacrylate,ethylene glycol dipropenoate,unii-42q902riye
75282
2-prop-2-enoyloxyethyl prop-2-enoate
C=CC(=O)OCCOC(=O)C=C
2274-11-5
C8H10O4
170.16
MFCD00010307
KUDUQBURMYMBIJ-UHFFFAOYSA-N
ethylene diacrylate,ethylene acrylate,ethylene glycol diacrylate,acrylic acid, ethylene ester,acrylic acid, ethylene glycol diester,2-propenoic acid, 1,2-ethanediyl ester,acrylic acid, diester with ethylene glycol,1,2-ethanediyl diacrylate,ethylene glycol dipropenoate,unii-42q902riye
75282
2-prop-2-enoyloxyethyl prop-2-enoate
C=CC(=O)OCCOC(=O)C=C
Melting Point 36°C to 40°C
Quantity 1g
Boiling Point 168°C to 169°C
Flash Point 73°C (163°F)
Beilstein 1862513
Formula Weight 165
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-2-fluorobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning