Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dichloro-2-fluorobenzene, 98%

Click to view available options
Quantity:
1g
5g
25g
2268-05-5
C6H3Cl2F
164.99
MFCD00010307
JORVCRLRRRRLFI-UHFFFAOYSA-N
2,6-dichlorofluorobenzene,1,3-dichloro-2-fluoro-benzene,2,6-dichloro-1-fluorobenzene,2,6-dichloro fluoro benzene,benzene,1,3-dichloro-2-fluoro,benzene, 1,3-dichloro-2-fluoro,pubchem2311,2,6-dichlorfluorbenzol,acmc-2097ae,2,6-dichloro fluorobenzene
137520
1,3-dichloro-2-fluorobenzene
C1=CC(=C(C(=C1)Cl)F)Cl
2268-05-5
C6H3Cl2F
164.99
MFCD00010307
JORVCRLRRRRLFI-UHFFFAOYSA-N
2,6-dichlorofluorobenzene,1,3-dichloro-2-fluoro-benzene,2,6-dichloro-1-fluorobenzene,2,6-dichloro fluoro benzene,benzene,1,3-dichloro-2-fluoro,benzene, 1,3-dichloro-2-fluoro,pubchem2311,2,6-dichlorfluorbenzol,acmc-2097ae,2,6-dichloro fluorobenzene
137520
1,3-dichloro-2-fluorobenzene
C1=CC(=C(C(=C1)Cl)F)Cl
Melting Point 36°C to 40°C
Quantity 25g
Boiling Point 168°C to 169°C
Flash Point 73°C (163°F)
Beilstein 1862513
Formula Weight 165
Percent Purity 98%
Chemical Name or Material 1,3-Dichloro-2-fluorobenzene

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning