Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dichloro-4-fluorobenzene, 99%

Click to view available options
Quantity:
50g
250g
1435-48-9
C6H3Cl2F
164.988
MFCD00000580
BDJZCCWUSOZUQG-UHFFFAOYSA-N
1,3-dichloro-4-fluorobenzene,2,4-dichlorofluorobenzene,benzene, 2,4-dichloro-1-fluoro,dcfb,benzene,2,4-dichloro-1-fluoro,2,4-dichloro-1-fluoro-benzene,pubchem2312,acmc-209cqu,2,4-dichlorofluobenzene,1,3-dichlorofluorobenzene
123112
2,4-dichloro-1-fluorobenzene
C1=CC(=C(C=C1Cl)Cl)F
1435-48-9
C6H3Cl2F
164.988
MFCD00000580
BDJZCCWUSOZUQG-UHFFFAOYSA-N
1,3-dichloro-4-fluorobenzene,2,4-dichlorofluorobenzene,benzene, 2,4-dichloro-1-fluoro,dcfb,benzene,2,4-dichloro-1-fluoro,2,4-dichloro-1-fluoro-benzene,pubchem2312,acmc-209cqu,2,4-dichlorofluobenzene,1,3-dichlorofluorobenzene
123112
2,4-dichloro-1-fluorobenzene
C1=CC(=C(C=C1Cl)Cl)F
Density 1.409
Refractive Index 1.525
Flash Point 79°C (174°F)
Quantity 250g
Boiling Point 172°C to 174°C
Melting Point -23°C
UN Number UN2810
Beilstein 2500791
Formula Weight 165
Percent Purity 99%
Chemical Name or Material 1,3-Dichloro-4-fluorobenzene
Show More Show Less

GHS H Statement
H373-H302-H315-H227
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
Combustible liquid.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P280-P301+P312-P303+P361+P353-P305+P351+P338-P314-P330-P332+P313-P363-P370+P378q-P501c

H226-H302-H315-H319-H373

DOTInformation : Hazard Class: 3; Packaging Group: III

EINECSNumber : 406-160-1

TSCA : No

Recommended Storage : Ambient temperatures