Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloro-5,5-dimethylhydantoin, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
118-52-5
C5H6Cl2N2O2
197.02
MFCD00003190
KEQGZUUPPQEDPF-UHFFFAOYSA-N
1,3-dichloro-5,5-dimethylhydantoin,dantoin,dactin,daktin,dichlorantin,omchlor,halane,hydan,hydantoin, dichlorodimethyl,hydan antiseptic
8360
1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
CC1(C(=O)N(C(=O)N1Cl)Cl)C
118-52-5
C5H6Cl2N2O2
197.02
MFCD00003190
KEQGZUUPPQEDPF-UHFFFAOYSA-N
1,3-dichloro-5,5-dimethylhydantoin,dantoin,dactin,daktin,dichlorantin,omchlor,halane,hydan,hydantoin, dichlorodimethyl,hydan antiseptic
8360
1,3-dichloro-5,5-dimethylimidazolidine-2,4-dione
CC1(C(=O)N(C(=O)N1Cl)Cl)C
Sublimation Point 100.0 °C
Color White
Flash Point 171°C
Insoluble Matter 1% max. (in chloroform)
Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 131°C to 136°C
Quantity 5g
Beilstein 24, II, 158
Merck Index 15, 3074
Solubility Information Solubility in water: 0.21% (25°C). Other solubilities: slowly forming hypochloric acid,9.2% in benzene- 30% in methylene chloride,32% in ethylene dichloride- 14% in chloroform,12.5% in ccl4- 17% in tetr
Formula Weight 197.02
Physical Form Crystalline Powder
Percent Purity ≥66% (available chlorine)
Chemical Name or Material 1, 3-Dichloro-5, 5-dimethylhydantoin
Show More Show Less

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
May intensify fire; oxidizer.
May cause an allergic skin reaction.
Very toxic to aquatic life.
Contact with acids liberates toxic gas.<br/

GHS P Statement
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger

EINECSNumber : 204-258-7

RTECSNumber : MU0700000

TSCA : TSCA