Alfa Aesar™ 1,3-Dichloro-5-nitrobenzene, 98%

Manufacturer: Alfa Aesar™ B2558414

Quantity 25g

 View more versions of this product

Catalog No. AAB2558414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 618-62-2
    Molecular Formula C6H3Cl2NO2
    Formula Weight 192
    MDL Number MFCD00007211
    UN Number UN3077
    Beilstein 2208877
    Synonym 3,5-Dichloronitrobenzene; 5-Nitro-m-dichlorobenzene
    Chemical Name or Material 1,3-Dichloro-5-nitrobenzene

    Specifications

    Formula Weight 192
    TSCA TSCA
    Molecular Formula C6H3Cl2NO2
    MDL Number MFCD00007211
    UN Number UN3077
    Beilstein 2208877
    Synonym 3,5-Dichloronitrobenzene; 5-Nitro-m-dichlorobenzene
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Quantity 25g
    CAS 618-62-2
    Melting Point 62° to 65°C
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H373-H302
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Health Hazard 3 GHS P Statement
    P280f-P273
    Wear protective clothing.
    Avoid release to the environment.
    Chemical Name or Material 1,3-Dichloro-5-nitrobenzene
    Assay 98%

    GHS H Statement
    H373-H302
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.

    GHS P Statement
    P280f-P273
    Wear protective clothing.
    Avoid release to the environment.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit