Alfa Aesar™ 1,3-Dichloroacetone, typically 99%

Manufacturer: Alfa Aesar™ 2230206

Quantity 5g

 View more versions of this product

Catalog No. AA2230206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 534-07-6
    Formula Weight 126.97
    MDL Number MFCD00000937
    UN Number UN2649
    Beilstein 605456
    Merck Index 14,3052
    Chemical Name or Material 1,3-Dichloroacetone

    Specifications

    Formula Weight 126.97
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    UN Number UN2649
    Boiling Point 173°C
    Beilstein 605456
    Melting Point 43° to 45°C
    Density 1.3834g/mL at 46°C
    TSCA TSCA
    Refractive Index 1.4400
    Physical Form Crystalline
    MDL Number MFCD00000937
    CAS 534-07-6
    Flash Point 89° (193°F)
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: 1,3-DICHLOROACETONE
    Merck Index 14,3052
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H314-H318-H341
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing genetic defects.
    RTECS Number UC1430000
    Quantity 5g
    Chemical Name or Material 1,3-Dichloroacetone
    Assay 99% (typically)

    GHS H Statement
    H300-H310-H330-H314-H318-H341
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Suspected of causing genetic defects.

    GHS P Statement
    P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit