Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichlorobenzene 98%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
250mL
500mL
1L
Packaging:
Glass bottle
541-73-1
C6H4Cl2
147
MFCD00000573
ZPQOPVIELGIULI-UHFFFAOYSA-N
m-dichlorobenzene,m-phenylene dichloride,m-dichlorobenzol,meta-dichlorobenzene,benzene, 1,3-dichloro,metadichlorobenzene,benzene, m-dichloro,m-phenylenedichloride,m-dcb,3-dichlorobenzene
10943
CHEBI:36693
1,3-dichlorobenzene
C1=CC(=CC(=C1)Cl)Cl
541-73-1
C6H4Cl2
147
MFCD00000573
ZPQOPVIELGIULI-UHFFFAOYSA-N
m-dichlorobenzene,m-phenylene dichloride,m-dichlorobenzol,meta-dichlorobenzene,benzene, 1,3-dichloro,metadichlorobenzene,benzene, m-dichloro,m-phenylenedichloride,m-dcb,3-dichlorobenzene
10943
CHEBI:36693
1,3-dichlorobenzene
C1=CC(=CC(=C1)Cl)Cl
Packaging Glass bottle
Refractive Index 1.5450 to 1.547
Melting Point -24°C
Flash Point 67°C
Boiling Point 172°C to 173°C
Specific Gravity 1.28
Viscosity 1.045 mPa.s (23°C)
Infrared Spectrum Authentic
Water 0.1% max.
Color Colorless to Yellow
Density 1.2800g/mL
Quantity 25mL
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H4Cl2
Beilstein 05, 202
Merck Index 15, 3064
Solubility Information Solubility in water: 0.0123g/100mL water (25°C). Other solubilities: soluble in organic solvents such as alcohol,,benzene,ether
Formula Weight 147
Physical Form Liquid
Percent Purity ≥97.5%
Chemical Name or Material 1, 3-Dichlorobenzene, 98%
Show More Show Less

GHS H Statement:
Toxic to aquatic life with long lasting effects.
Harmful if swallowed.

GHS P Statement:
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 208-792-1

RTECSNumber : CZ4499000

TSCA : TSCA