Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dichloroisoquinoline, 97%, Acros Organics

Click to view available options
Quantity:
1g
5g
7742-73-6
C9H5Cl2N
198.05
BRGZEQXWZWBPJH-UHFFFAOYSA-N
isoquinoline, 1,3-dichloro,1,3-dichloro-isoquinoline,pubchem7254,1, 3-dichloroisoquinoline,acmc-209p9x,ksc493q3b,1,3-dichloroisoquinoline
298625
1,3-dichloroisoquinoline
C1=CC=C2C(=C1)C=C(N=C2Cl)Cl
7742-73-6
C9H5Cl2N
198.05
BRGZEQXWZWBPJH-UHFFFAOYSA-N
isoquinoline, 1,3-dichloro,1,3-dichloro-isoquinoline,pubchem7254,1, 3-dichloroisoquinoline,acmc-209p9x,ksc493q3b,1,3-dichloroisoquinoline
298625
1,3-dichloroisoquinoline
C1=CC=C2C(=C1)C=C(N=C2Cl)Cl
Melting Point 120.0°C to 122.0°C
Packaging Glass bottle
Quantity 1g
Assay Percent Range 97%
Formula Weight 198.05
Percent Purity 97%
Chemical Name or Material 1, 3-Dichloroisoquinoline