Alfa Aesar™ 1,3-Diethoxybenzene, 95%

Manufacturer: Alfa Aesar™ A1133830

Quantity 250g

 View more versions of this product

Catalog No. AAA1133830

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2049-73-2
    Molecular Formula C10H14O2
    Formula Weight 166.22
    MDL Number MFCD00015145
    Beilstein 2517730
    Synonym Resorcinol diethyl ether
    Chemical Name or Material 1,3-Diethoxybenzene

    Specifications

    Formula Weight 166.22
    CAS 2049-73-2
    Boiling Point 234° to 236°C
    Quantity 250g
    Molecular Formula C10H14O2
    Beilstein 2517730
    Melting Point 11° to 12°C
    MDL Number MFCD00015145
    Synonym Resorcinol diethyl ether
    Chemical Name or Material 1,3-Diethoxybenzene
    Assay 95%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit