Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diethoxybenzene, 95%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
2049-73-2
C10H14O2
166.22
MFCD00015145
MKGFYMKFBCWNCP-UHFFFAOYSA-N
m-diethoxybenzene,benzene, 1,3-diethoxy,resorcinol diethyl ether,resorcindiathylather,benzene,3-diethoxy,acmc-1cdp4,benzene, 1,3-diethoxy-9ci
74899
1,3-diethoxybenzene
CCOC1=CC(=CC=C1)OCC
2049-73-2
C10H14O2
166.22
MFCD00015145
MKGFYMKFBCWNCP-UHFFFAOYSA-N
m-diethoxybenzene,benzene, 1,3-diethoxy,resorcinol diethyl ether,resorcindiathylather,benzene,3-diethoxy,acmc-1cdp4,benzene, 1,3-diethoxy-9ci
74899
1,3-diethoxybenzene
CCOC1=CC(=CC=C1)OCC
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Refractive Index 1.5080 to 1.5100 (20°C, 589nm)
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Linear Formula C6H4(OC2H5)2
Formula Weight 166.22
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material 1, 3-Diethoxybenzene, 95%
Show More Show Less