Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diethyl-2-thiobarbituric acid, 99%, ACROS Organics™

Click to view available options
Quantity:
50g
250g
Packaging:
Glass bottle
5217-47-0
C8H12N2O2S
200.256
MFCD00006676
SHBTUGJAKBRBBJ-UHFFFAOYSA-N
1,3-diethyl-2-thiobarbituric acid,1,3-diethylthiobarbituric acid,n,n'-diethylthiobarbituric acid,n,n'-diethyl-2-thiobarbituric acid,1,3-diethyl-2-thioxodihydropyrimidine-4,6 1h,5h-dione,unii-bxb40m6a7w,4,6 1h,5h-pyrimidinedione, 1,3-diethyldihydro-2-thioxo,barbituric acid, 1,3-diethyl-2-thio,bxb40m6a7w,1,3-diethyldihydro-2-thioxopyrimidine-4,6 1h,5h-dione
78890
1,3-diethyl-2-sulfanylidene-1,3-diazinane-4,6-dione
CCN1C(=O)CC(=O)N(C1=S)CC
5217-47-0
C8H12N2O2S
200.256
MFCD00006676
SHBTUGJAKBRBBJ-UHFFFAOYSA-N
1,3-diethyl-2-thiobarbituric acid,1,3-diethylthiobarbituric acid,n,n'-diethylthiobarbituric acid,n,n'-diethyl-2-thiobarbituric acid,1,3-diethyl-2-thioxodihydropyrimidine-4,6 1h,5h-dione,unii-bxb40m6a7w,4,6 1h,5h-pyrimidinedione, 1,3-diethyldihydro-2-thioxo,barbituric acid, 1,3-diethyl-2-thio,bxb40m6a7w,1,3-diethyldihydro-2-thioxopyrimidine-4,6 1h,5h-dione
78890
1,3-diethyl-2-sulfanylidene-1,3-diazinane-4,6-dione
CCN1C(=O)CC(=O)N(C1=S)CC
Melting Point 105°C to 109°C
Color White to Yellow
Quantity 50g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Beilstein 24, IV, 1885
Formula Weight 200.25
Physical Form Powder or Crystals
Percent Purity ≥98.5%
Chemical Name or Material 1, 3-Diethyl-2-thiobarbituric acid
Show More Show Less