Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Difluoro-2-propanol, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
10g
Packaging:
Glass Bottle
453-13-4
C3H6F2O
96.077
MFCD00000452
PVDLUGWWIOGCNH-UHFFFAOYSA-N
1,3-difluoro-2-propanol,2-propanol, 1,3-difluoro,1,3-difluoroisopropanol,2-propanol,3-difluoro,wln: f1yq1f,sgqdlibrbrwup@,acmc-20ap4b,1,3-difluoropropane-2-ol,1,3-difluoro-propan-2-ol,1,3-difluoro-2-hydroxypropane
67985
1,3-difluoropropan-2-ol
C(C(CF)O)F
453-13-4
C3H6F2O
96.077
MFCD00000452
PVDLUGWWIOGCNH-UHFFFAOYSA-N
1,3-difluoro-2-propanol,2-propanol, 1,3-difluoro,1,3-difluoroisopropanol,2-propanol,3-difluoro,wln: f1yq1f,sgqdlibrbrwup@,acmc-20ap4b,1,3-difluoropropane-2-ol,1,3-difluoro-propan-2-ol,1,3-difluoro-2-hydroxypropane
67985
1,3-difluoropropan-2-ol
C(C(CF)O)F
Infrared Spectrum Authentic
Quantity 5g
Density 1.2400g/mL
Packaging Glass Bottle
Refractive Index 1.3720 to 1.3750 (20°C, 589nm)
Color Brown to Yellow
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula FCH2CH(OH)CH2F
Formula Weight 96.08
Physical Form Liquid
Percent Purity ≥97.5% (GC)
Chemical Name or Material 1, 3-Difluoro-2-propanol, 99%
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 207-216-6

RTECSNumber : UB1770000

TSCA : TSCA