Learn More
1,3-Dihydroxyacetone dimer, 97%, Thermo Scientific Chemicals
Description
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,3-Dihydroxyacetone dimer is used as a catalytic agent and petrochemical additive. It is also used as a building block in organic synthesis. Further, it serves as a reagent in three-carbon nucleophilic or electrophilic component in various reactions.
Solubility
Soluble in ethanol, ethyl ether, acetone. Slightly soluble in water. Insoluble in ligroin.
Notes
Hygroscopic. Store in a cool place. Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∼75°C to 82°C |
Quantity | 50 g |
Beilstein | 112910 |
Merck Index | 14,3182 |
Solubility Information | Soluble in ethanol,ethyl ether,acetone. Slightly soluble in water. Insoluble in ligroin. |
IUPAC Name | 2,5-bis(hydroxymethyl)-1,4-dioxane-2,5-diol |
Formula Weight | 180.16 |
Percent Purity | 97% |
Chemical Name or Material | 1,3-Dihydroxyacetone dimer |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 612-947-2 |
TSCA | No |
Recommended Storage | Keep cold; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.