Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Diisopropylbenzene, 96%

Click to view available options
Quantity:
50g
250g
99-62-7
C12H18
162.276
MFCD00008889
UNEATYXSUBPPKP-UHFFFAOYSA-N
1,3-diisopropylbenzene,m-diisopropylbenzene,benzene, 1,3-bis 1-methylethyl,benzene, m-diisopropyl,m-diisopropylbenzol,1,3-bis 1-methylethyl benzene,3-isopropylcumene,unii-482vxw192e,meta-diisopropylbenzene,chembl31352
7450
1,3-di(propan-2-yl)benzene
CC(C)C1=CC(=CC=C1)C(C)C
99-62-7
C12H18
162.276
MFCD00008889
UNEATYXSUBPPKP-UHFFFAOYSA-N
1,3-diisopropylbenzene,m-diisopropylbenzene,benzene, 1,3-bis 1-methylethyl,benzene, m-diisopropyl,m-diisopropylbenzol,1,3-bis 1-methylethyl benzene,3-isopropylcumene,unii-482vxw192e,meta-diisopropylbenzene,chembl31352
7450
1,3-di(propan-2-yl)benzene
CC(C)C1=CC(=CC=C1)C(C)C
Applications
1,3-Diisopropylbenzene was used in the synthesis of benzene-1,3,5-triphosphonic acid.

Solubility
Soluble in water 4.33 mg/L at 25°C.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
Flash Point 76°C (170°F)
Melting Point -63°C
Quantity 50g
Boiling Point 204°C
Beilstein 1905828
Formula Weight 162.27
Percent Purity 96%
Chemical Name or Material 1,3-Diisopropylbenzene

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : CZ6334000

TSCA : TSCA