Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diisopropylimidazolium chloride, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
139143-09-2
C9H17ClN2
188.699
MFCD03840577
DOFXKPAOJLLPII-UHFFFAOYSA-M
1,3-diisopropylimidazolium chloride,1,3-di-i-propylimidazolium chloride,1,3-diisopropyl-1h-imidazol-3-ium chloride,1,3-diisopropylimidazol-1-ium chloride,diic,acmc-20ap45,n,n'-isopropyl imidazolium chloride,1h-imidazolium,1,3-bis 1-methylethyl-, chloride 1:1,n,n inverted exclamation marka-isopropyl imidazolium chloride
11229242
1,3-di(propan-2-yl)imidazol-1-ium;chloride
CC(C)N1C=C[N+](=C1)C(C)C.[Cl-]
139143-09-2
C9H17ClN2
188.699
MFCD03840577
DOFXKPAOJLLPII-UHFFFAOYSA-M
1,3-diisopropylimidazolium chloride,1,3-di-i-propylimidazolium chloride,1,3-diisopropyl-1h-imidazol-3-ium chloride,1,3-diisopropylimidazol-1-ium chloride,diic,acmc-20ap45,n,n'-isopropyl imidazolium chloride,1h-imidazolium,1,3-bis 1-methylethyl-, chloride 1:1,n,n inverted exclamation marka-isopropyl imidazolium chloride
11229242
1,3-di(propan-2-yl)imidazol-1-ium;chloride
CC(C)N1C=C[N+](=C1)C(C)C.[Cl-]
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color White to Beige
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 188.7
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 1, 3-Diisopropylimidazolium chloride
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.