Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethyl-2-imidazolidinone, 98%, ACROS Organics™

Click to view available options
Quantity:
100g
1kg
Packaging:
Glass bottle
80-73-9
C5H10N2O
114.15
MFCD00003188
CYSGHNMQYZDMIA-UHFFFAOYSA-N
1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
6661
1,3-dimethylimidazolidin-2-one
CN1CCN(C1=O)C
80-73-9
C5H10N2O
114.15
MFCD00003188
CYSGHNMQYZDMIA-UHFFFAOYSA-N
1,3-dimethyl-2-imidazolidinone,n,n'-dimethylethyleneurea,dimethyl imidazolidinone,n,n'-dimethylimidazolidinone,1,3-dimethylimidazolidone,rhonite 1,dmeu,karbomos tsem,2-imidazolidinone, 1,3-dimethyl,1,3-dimethylethyleneurea
6661
1,3-dimethylimidazolidin-2-one
CN1CCN(C1=O)C
Viscosity 1.944 mPa.s (25°C)
Melting Point 8.2°C
Water 0.1% max.
Specific Gravity 1.05
Density 1.0500g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4710 to 1.4730
Boiling Point 225.5°C
Flash Point 93°C
Quantity 100g
Color Undesignated
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Fieser 11,202; 15,145
Merck Index 15, 3273
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in methanol, ethanol, 2-propanol, ether,, methylene chloride, dmf, acetone, ketone and, chloroform
Formula Weight 114.15
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material 1,3-Dimethyl-2-imidazolidinone, 98%
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with

GHS Signal Word: Danger

EINECSNumber : 201-304-8

RTECSNumber : NJ0660000

TSCA : TSCA

Recommended Storage : may discolor during storage