Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dimethyladamantane, 98%

Click to view available options
Quantity:
1g
5g
25g
702-79-4
C12H20
164.292
MFCD00074755
CWNOIUTVJRWADX-UHFFFAOYSA-N
adamantane, 1,3-dimethyl,1-3-dimethyladamantane,1,3-dimethyl adamantane,unii-96g4od5z59,1,3-dimethyltricyclo 3.3.1.13,7 decane,1,3-dimethyltricyclo 3.3.1.1 3,7 decane,tricyclo 3.3.1.1 3,7 decane, 1,3-dimethyl,1r,3r,5r,7r-1,3-dimethyladamantane,memantine intermediate
12800
CHEBI:47900
1,3-dimethyladamantane
CC12CC3CC(C1)CC(C3)(C2)C
702-79-4
C12H20
164.292
MFCD00074755
CWNOIUTVJRWADX-UHFFFAOYSA-N
adamantane, 1,3-dimethyl,1-3-dimethyladamantane,1,3-dimethyl adamantane,unii-96g4od5z59,1,3-dimethyltricyclo 3.3.1.13,7 decane,1,3-dimethyltricyclo 3.3.1.1 3,7 decane,tricyclo 3.3.1.1 3,7 decane, 1,3-dimethyl,1r,3r,5r,7r-1,3-dimethyladamantane,memantine intermediate
12800
CHEBI:47900
1,3-dimethyladamantane
CC12CC3CC(C1)CC(C3)(C2)C
Density 0.902
Refractive Index 1.478
Boiling Point 201°C
Quantity 1g
Flash Point 52°C (126°F)
UN Number UN3295
Formula Weight 164.29
Percent Purity 98%
Chemical Name or Material 1,3-Dimethyladamantane

GHS H Statement
H226
Flammable liquid and vapour.

P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c

H226

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.

EINECSNumber : 211-870-8

TSCA : No

Recommended Storage : Ambient temperatures