Alfa Aesar™ 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 98%

Manufacturer: Alfa Aesar™ B2505722

Quantity 100g

 View more versions of this product

Catalog No. AAB2505722

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1892-57-5
    Molecular Formula C8H17N3
    Linear Formula CH3CH2N=C=N(CH2)3N(CH3)2
    Formula Weight 155.24
    MDL Number MFCD00044916
    UN Number UN2735
    Beilstein 507429
    Synonym N-Ethyl-N'-(3-dimethylaminopropyl)carbodiimide
    Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide

    Specifications

    Formula Weight 155.24
    TSCA TSCA
    Flash Point >110°C (230°F)
    Synonym N-Ethyl-N'-(3-dimethylaminopropyl)carbodiimide
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
    Molecular Formula C8H17N3
    Linear Formula CH3CH2N=C=N(CH2)3N(CH3)2
    Boiling Point 66° to 68°C (1mmHg)
    MDL Number MFCD00044916
    CAS 1892-57-5
    UN Number UN2735
    Beilstein 507429
    Quantity 100g
    Health Hazard 1 Danger
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Recommended Storage −30° to −10°C
    Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide
    Assay 98%

    Description

    Applications
    1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide is used as a carboxyl activating agent and activate phosphate groups in phospho mono and di esters. It is used in peptide synthesis, 3 ft.-amino-3 ft.-deoxyadenosine-5 ft.-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

    Solubility
    Soluble in water.

    Notes
    Air sensitive. Incompatible with strong acids and strong oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    −30° to −10°C

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit