Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide, 98%

Click to view available options
Quantity:
5g
25g
100g
1892-57-5
C8H17N3
155.245
MFCD00044916
LMDZBCPBFSXMTL-UHFFFAOYSA-N
1-3-dimethylaminopropyl-3-ethylcarbodiimide,edac,1-ethyl-3-3-dimethylaminopropyl carbodiimide,n1-ethylimino methylene-n3,n3-dimethylpropane-1,3-diamine,unii-rj5ozg6i4a,edci,1,3-propanediamine, n'-ethylcarbonimidoyl-n,n-dimethyl,n-3-dimethylaminopropyl-n'-ethylcarbodiimide,rj5ozg6i4a,ethyldimethylaminopropyl carbodiimide
15908
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine
CCN=C=NCCCN(C)C
1892-57-5
C8H17N3
155.245
MFCD00044916
LMDZBCPBFSXMTL-UHFFFAOYSA-N
1-3-dimethylaminopropyl-3-ethylcarbodiimide,edac,1-ethyl-3-3-dimethylaminopropyl carbodiimide,n1-ethylimino methylene-n3,n3-dimethylpropane-1,3-diamine,unii-rj5ozg6i4a,edci,1,3-propanediamine, n'-ethylcarbonimidoyl-n,n-dimethyl,n-3-dimethylaminopropyl-n'-ethylcarbodiimide,rj5ozg6i4a,ethyldimethylaminopropyl carbodiimide
15908
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine
CCN=C=NCCCN(C)C
Applications
1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide is used as a carboxyl activating agent and activate phosphate groups in phospho mono and di esters. It is used in peptide synthesis, 3′-amino-3′-deoxyadenosine-5′-di- and triphosphates and in the preparation of antibodies like immunoconjugates. It plays a vital role for immobilization of large biomolecules in association with N-hydroxysuccinimide. It is also used in the acylation of phosphoranes.

Solubility
Soluble in water.

Notes
Air sensitive. Incompatible with strong acids and strong oxidizing agents.
Refractive Index 1.461
Density 0.885
Sensitivity Air and moisture sensitive
Quantity 25g
Flash Point >110°C (230°F)
Boiling Point 66°C to 68°C (1mmHg)
Linear Formula CH3CH2N=C=N(CH2)3N(CH3)2
UN Number UN2735
Beilstein 507429
Solubility Information Soluble in water.
Formula Weight 155.24
Percent Purity 98%
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide
Show More Show Less

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

EINECSNumber : 217-579-2

TSCA : Yes

Recommended Storage : Store at -20°C; Store under Argon