Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide hydrochloride, 98+%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
50g
100g
500g
Packaging:
Glass Bottle
25952-53-8
C8H17N3·HCl
191.71
MFCD00012503
FPQQSJJWHUJYPU-UHFFFAOYSA-N
1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride,edc.hcl,edci,edc hydrochloride,wsc hcl,edac hydrochloride,edac, hydrochloride,1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride,edcl,edac hcl
2723939
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride
CCN=C=NCCCN(C)C.Cl
25952-53-8
C8H17N3·HCl
191.71
MFCD00012503
FPQQSJJWHUJYPU-UHFFFAOYSA-N
1-3-dimethylaminopropyl-3-ethylcarbodiimide hydrochloride,edc.hcl,edci,edc hydrochloride,wsc hcl,edac hydrochloride,edac, hydrochloride,1-ethyl-3-3-dimethylaminopropyl carbodiimide hydrochloride,edcl,edac hcl
2723939
3-(ethyliminomethylideneamino)-N,N-dimethylpropan-1-amine;hydrochloride
CCN=C=NCCCN(C)C.Cl
Coupling reagent
Packaging Glass Bottle
Infrared Spectrum Authentic
Color White
Melting Point 111.0°C to 116.0°C
Quantity 100g
CAS Min % 98.0
CAS Max % 100.0
Assay Percent Range 98% min. (Argentometry)
Linear Formula (CH3)2NCH2CH2CH2N=C=NCH2CH3
UN Number 2811
Fieser 12,199
Merck Index 15, 3555
Solubility Information Solubility in water: >200g/l. Other solubilities: soluble in ethanol
Formula Weight 191.71
Physical Form Crystalline Powder
Percent Purity 98+%
Chemical Name or Material 1-(3-Dimethylaminopropyl)-3-ethylcarbodiimide Hydrochloride
Show More Show Less

GHS H Statement
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical

GHS Signal Word: Warning

EINECSNumber : 247-361-2