Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Dimethylaminopropyl)piperazine, 99%, Thermo Scientific™

Catalog Number aah5069003
Click to view available options
Quantity:
1 g
877-96-3
C9H21N3
171.288
MFCD00082601
YJRGRZJKGMBHIB-UHFFFAOYSA-N
1-3-dimethylaminopropyl piperazine, n,n-dimethyl-3-piperazin-1-yl propan-1-amine, dimethyl 3-piperazin-1-yl propyl amine, 1-3-dimethylaminopropyl-piperazine, 1-3-dimethylamino propyl piperazine, 1-3-dimethylamino-propyl-piperazine, 1-piperazinepropanamine,n,n-dimethyl, dimethyl-3-piperazin-1-yl-propyl-amine, n,n-dimethyl-3-1-piperazinyl-1-propanamine, n,n-dimethyl-n-3-1-piperazinyl propyl amine
2736234
N,N-dimethyl-3-piperazin-1-ylpropan-1-amine
CN(C)CCCN1CCNCC1
877-96-3
C9H21N3
171.288
MFCD00082601
YJRGRZJKGMBHIB-UHFFFAOYSA-N
1-3-dimethylaminopropyl piperazine, n,n-dimethyl-3-piperazin-1-yl propan-1-amine, dimethyl 3-piperazin-1-yl propyl amine, 1-3-dimethylaminopropyl-piperazine, 1-3-dimethylamino propyl piperazine, 1-3-dimethylamino-propyl-piperazine, 1-piperazinepropanamine,n,n-dimethyl, dimethyl-3-piperazin-1-yl-propyl-amine, n,n-dimethyl-3-1-piperazinyl-1-propanamine, n,n-dimethyl-n-3-1-piperazinyl propyl amine
2736234
N,N-dimethyl-3-piperazin-1-ylpropan-1-amine
CN(C)CCCN1CCNCC1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Formula Weight 171.29
Percent Purity 99%
Chemical Name or Material 1-(3-Dimethylaminopropyl)piperazine
Hazard Category H312-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit