Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dimethylcyclohexane, 97%, mixture of cis and trans, Acros Organics

Click to view available options
Quantity:
5mL
25mL
591-21-9
C8H16
112.216
SGVUHPSBDNVHKL-UHFFFAOYSA-N
hexahydro-m-xylene,cyclohexane, 1,3-dimethyl,1,3-dimethylcyclohexane,c&t,trans-1,3-dimethylcyclohexane,1,3-dimethylcyclohexane, cis + trans,unii-9sgo12qv70,1,3-dimethylcyclohexane, cis,m-dimethylcyclohexane,acmc-209nia,1,3-dimethyl cyclohexane
11564
1,3-dimethylcyclohexane
CC1CCCC(C1)C
591-21-9
C8H16
112.216
SGVUHPSBDNVHKL-UHFFFAOYSA-N
hexahydro-m-xylene,cyclohexane, 1,3-dimethyl,1,3-dimethylcyclohexane,c&t,trans-1,3-dimethylcyclohexane,1,3-dimethylcyclohexane, cis + trans,unii-9sgo12qv70,1,3-dimethylcyclohexane, cis,m-dimethylcyclohexane,acmc-209nia,1,3-dimethyl cyclohexane
11564
1,3-dimethylcyclohexane
CC1CCCC(C1)C
Quantity 25mL
Infrared Spectrum Authentic
Refractive Index 1.4260 to 1.4310 (20°C, 589nm)
Color Colorless
Density 0.77g/mL
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Linear Formula C6H10(CH3)
Formula Weight 112.21
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 1, 3-Dimethylcyclohexane, 97%
Show More Show Less

EINECSNumber : 209-707-0

TSCA : TSCA