Alfa Aesar™ 1,3-Dimethylimidazolium dimethyl phosphate, 98%

Manufacturer: Alfa Aesar™ H2778818

Quantity 50g

 View more versions of this product

Catalog No. AAH2778818

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 654058-04-5
    Molecular Formula C7H15N2O4P
    Formula Weight 222.18
    MDL Number MFCD08275369
    Chemical Name or Material 1,3-Dimethylimidazolium dimethyl phosphate

    Specifications

    Formula Weight 222.18
    Molecular Formula C7H15N2O4P
    MDL Number MFCD08275369
    CAS 654058-04-5
    Quantity 50g
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 1,3-Dimethylimidazolium dimethyl phosphate
    Assay 98%

    Description

    Applications
    It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. ionic liquid 1,3-dimethylimidazolium dimethylphosphate ([MMIM][DMP]) + water/ethanol/methanol mixtures exhibit properties which render them suitable as candidates for working pairs in industrial applications of absorption heat pumps or chillers.

    Solubility
    Insoluble in water.

    Notes
    Store in cool dry conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit