Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Dimethylorotic acid, 97%

Click to view available options
Quantity:
1g
5g
4116-38-5
C7H8N2O4
184.151
MFCD00090746
ZXDYFTWNWCMEOS-UHFFFAOYSA-N
1,3-dimethylorotic acid,1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid,1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid,1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-pyrimidine-4-carboxylic acid,4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-1,3-dimethyl-2,6-dioxo,1,3-dimethyloroticacid,1,3-dimethyl-2,6-dioxo-1,3-dihydropyrimidine-4-carboxylic acid,4-pyrimidinecarboxylicacid, 1,2,3,6-tetrahydro-1,3-dimethyl-2,6-dioxo
5025211
1,3-dimethyl-2,6-dioxopyrimidine-4-carboxylic acid
CN1C(=CC(=O)N(C1=O)C)C(=O)O
4116-38-5
C7H8N2O4
184.151
MFCD00090746
ZXDYFTWNWCMEOS-UHFFFAOYSA-N
1,3-dimethylorotic acid,1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylic acid,1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-4-pyrimidinecarboxylic acid,1,3-dimethyl-2,6-dioxo-1,2,3,6-tetrahydro-pyrimidine-4-carboxylic acid,4-pyrimidinecarboxylic acid, 1,2,3,6-tetrahydro-1,3-dimethyl-2,6-dioxo,1,3-dimethyloroticacid,1,3-dimethyl-2,6-dioxo-1,3-dihydropyrimidine-4-carboxylic acid,4-pyrimidinecarboxylicacid, 1,2,3,6-tetrahydro-1,3-dimethyl-2,6-dioxo
5025211
1,3-dimethyl-2,6-dioxopyrimidine-4-carboxylic acid
CN1C(=CC(=O)N(C1=O)C)C(=O)O
Quantity 1g
Beilstein 188093
Formula Weight 184.15
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 1,3-Dimethylorotic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning