Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Dioxolane, ReagentPlus™, 99%, Honeywell™

Click to view available options
Quantity:
1L
4L
18L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
646-06-0
C3H6O2
74.079
MFCD00003207
WNXJIVFYUVYPPR-UHFFFAOYSA-N
dioxolane,glycolformal,formal glycol,1,3-dioxolan,1,3-dioxacyclopentane,1,3-dioxalane,ethylene glycol formal,glycol methylene ether,glycol formal,dioxolan czech
12586
CHEBI:87597
1,3-dioxolane
C1COCO1
646-06-0
C3H6O2
74.079
MFCD00003207
WNXJIVFYUVYPPR-UHFFFAOYSA-N
dioxolane,glycolformal,formal glycol,1,3-dioxolan,1,3-dioxacyclopentane,1,3-dioxalane,ethylene glycol formal,glycol methylene ether,glycol formal,dioxolan czech
12586
CHEBI:87597
1,3-dioxolane
C1COCO1

Reagent Grade, contains ~75 ppm BHT as inhibitor, 99%

  • ≥95% for general laboratory use.
Density 1.06g/mL at 25°C
Color Colorless
Quantity 18L
Boiling Point 76°C
Ignition Point 525°F
Melting Point -95°C
Flash Point −3°C (26.6°F)
Packaging Stainless Steel Can
Vapor Pressure 70mmHg (20°C)
Refractive Index n20/D 1.401(lit.)
UN Number UN1166
Beilstein 102453
Formula Weight 74.08g/mol
Percent Purity 99%
Grade ReagentPlus™
Chemical Name or Material 1,3-Dioxolane, Contains approx.75 ppm BHT as inhibitor
Show More Show Less

P210-P305 + P351 + P338-P370 + P378-P403 + P235

H225-H319

DOTInformation : Class 3 - PG 2 - Dioxolane

EINECSNumber : 211-463-5

RTECSNumber : JH6760000