Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Diphenoxybenzene, 99%

Click to view available options
Quantity:
5g
25g
3379-38-2
C18H14O2
262.308
MFCD00003036
JTNRGGLCSLZOOQ-UHFFFAOYSA-N
m-diphenoxybenzene,benzene, 1,3-diphenoxy,m-diphenyloxybenzene,m-phenoxyphenoxybenzene,benzene, m-diphenoxy,resorcinol diphenyl ether,1,1'-1,3-phenylenebis oxy dibenzene,m-diphenoxybenzol,acmc-1adxk
76911
CHEBI:39273
1,3-diphenoxybenzene
C1=CC=C(C=C1)OC2=CC(=CC=C2)OC3=CC=CC=C3
3379-38-2
C18H14O2
262.308
MFCD00003036
JTNRGGLCSLZOOQ-UHFFFAOYSA-N
m-diphenoxybenzene,benzene, 1,3-diphenoxy,m-diphenyloxybenzene,m-phenoxyphenoxybenzene,benzene, m-diphenoxy,resorcinol diphenyl ether,1,1'-1,3-phenylenebis oxy dibenzene,m-diphenoxybenzol,acmc-1adxk
76911
CHEBI:39273
1,3-diphenoxybenzene
C1=CC=C(C=C1)OC2=CC(=CC=C2)OC3=CC=CC=C3
Quantity 5g
Melting Point 59°C to 61°C
Beilstein 2054179
Formula Weight 262.31
Percent Purity 99%
Chemical Name or Material 1,3-Diphenoxybenzene

EINECSNumber : 222-181-7

TSCA : Yes

Recommended Storage : Ambient temperatures