Alfa Aesar™ 1,3-Diphenylguanidine, primary standard, 99+%

Primary material for standardizing acids

Manufacturer: Alfa Aesar™ 3139518

Quantity 50g

 View more versions of this product

Catalog No. AA3139518

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 102-06-7
    Formula Weight 211.27
    MDL Number MFCD00001758
    UN Number UN3077
    Beilstein 1875653
    Merck Index 14,3324
    Synonym N,N'-Diphenylguanidine; Melaniline
    Chemical Name or Material 1,3-Diphenylguanidine, Primary standard

    Specifications

    Formula Weight 211.27
    Health Hazard 3 GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Characteristic
    MDL Number MFCD00001758
    UN Number UN3077
    RTECS Number MF0875000
    Physical Form Powder
    Beilstein 1875653
    Synonym N,N'-Diphenylguanidine; Melaniline
    Density 1.130g/mL
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    CAS 102-06-7
    Merck Index 14,3324
    Melting Point 148° to 150°C
    Boiling Point 170°C (dec.)
    Health Hazard 1 Warning
    TSCA TSCA
    Color Pale Cream
    Quantity 50g
    Health Hazard 2 GHS H Statement
    H361-H302-H315-H319-H335
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material 1,3-Diphenylguanidine, Primary standard
    Assay ≥99%

    GHS H Statement
    H361-H302-H315-H319-H335
    Suspected of damaging fertility or the unborn child.
    Harmful if swallowed.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P281-P305+P351+P338-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Use personal protective equipment as required.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit