Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3-Diphenylisobenzofuran, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
5471-63-6
C20H14O
270.32
MFCD00005931
ZKSVYBRJSMBDMV-UHFFFAOYSA-N
1,3-diphenylisobenzofuran,diphenylisobenzofuran,isobenzofuran, 1,3-diphenyl,1,3 diphenylisobenzofuran,2,5-diphenyl-3,4-benzofuran,acmc-209lj9,1,3-diphenylbenzo c furan,1,3-diphenylisobenzo b furan,1,3-di phenyl-2-benzofuran,#
21649
1,3-diphenyl-2-benzofuran
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4
5471-63-6
C20H14O
270.32
MFCD00005931
ZKSVYBRJSMBDMV-UHFFFAOYSA-N
1,3-diphenylisobenzofuran,diphenylisobenzofuran,isobenzofuran, 1,3-diphenyl,1,3 diphenylisobenzofuran,2,5-diphenyl-3,4-benzofuran,acmc-209lj9,1,3-diphenylbenzo c furan,1,3-diphenylisobenzo b furan,1,3-di phenyl-2-benzofuran,#
21649
1,3-diphenyl-2-benzofuran
C1=CC=C(C=C1)C2=C3C=CC=CC3=C(O2)C4=CC=CC=C4
Alkylation reagent
Color Yellow to Green
Packaging Glass bottle
Infrared Spectrum Authentic
Melting Point 129.0°C to 133.0°C
Quantity 5g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 17, 87
Fieser 01,342; 02,178; 04,209
Formula Weight 270.32
Physical Form Crystalline Solid or Needles
Percent Purity 97%
Chemical Name or Material 1, 3-Diphenylisobenzofuran, 99+%
Show More Show Less

AIR SENSITIVE

LIGHT SENSITIVE

EINECSNumber : 226-808-5

TSCA : TSCA