Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Fluorophenyl)-2-thiourea, 98%, Acros Organics

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
458-05-9
C7H7FN2S
170.21
YKOBLHQALWQKTC-UHFFFAOYSA-N
1-3-fluorophenyl-2-thiourea,1-3-fluorophenyl thiourea,3-fluorophenyl thiourea,n-3-fluorophenyl thiourea,amino 3-fluorophenyl amino methane-1-thione,acmc-20apbn,1-m-fluorophenyl thiourea,3-fluoro-phenyl-thiourea
2734799
(3-fluorophenyl)thiourea
C1=CC(=CC(=C1)F)NC(=S)N
458-05-9
C7H7FN2S
170.21
YKOBLHQALWQKTC-UHFFFAOYSA-N
1-3-fluorophenyl-2-thiourea,1-3-fluorophenyl thiourea,3-fluorophenyl thiourea,n-3-fluorophenyl thiourea,amino 3-fluorophenyl amino methane-1-thione,acmc-20apbn,1-m-fluorophenyl thiourea,3-fluoro-phenyl-thiourea
2734799
(3-fluorophenyl)thiourea
C1=CC(=CC(=C1)F)NC(=S)N
Melting Point 113°C to 117°C
Quantity 5g
Packaging Glass bottle
Assay Percent Range 97.5% min. (HPLC)
Linear Formula F6C6H4NHCSNH2
Solubility Information Solubility in water: insoluble
Formula Weight 170.21
Percent Purity 98%
Chemical Name or Material 1-(3-Fluorophenyl)-2-thiourea

GHS Signal Word: Danger