Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(3-Fluorophenyl)ethanol, 97%

Click to view available options
Quantity:
5g
25g
100g
402-63-1
C8H9FO
140.157
MFCD00014402
YESOPGLEIJQAEF-UHFFFAOYSA-N
1-3-fluorophenyl ethanol,1-3-fluorophenyl ethan-1-ol,3-fluoro-alpha-methylbenzyl alcohol,3-fluorophenylmethylcarbinol,benzenemethanol, 3-fluoro-.alpha.-methyl,benzenemethanol,3-fluoro-a-methyl-, ar,benzenemethanol,3-fluoro-a-methyl-, as,acmc-20dpsl,acmc-20a9u3,m-fluorophenylmethylcarbinol
95348
1-(3-fluorophenyl)ethanol
CC(C1=CC(=CC=C1)F)O
402-63-1
C8H9FO
140.157
MFCD00014402
YESOPGLEIJQAEF-UHFFFAOYSA-N
1-3-fluorophenyl ethanol,1-3-fluorophenyl ethan-1-ol,3-fluoro-alpha-methylbenzyl alcohol,3-fluorophenylmethylcarbinol,benzenemethanol, 3-fluoro-.alpha.-methyl,benzenemethanol,3-fluoro-a-methyl-, ar,benzenemethanol,3-fluoro-a-methyl-, as,acmc-20dpsl,acmc-20a9u3,m-fluorophenylmethylcarbinol
95348
1-(3-fluorophenyl)ethanol
CC(C1=CC(=CC=C1)F)O
Boiling Point 104°C to 106°C (20mmHg)
Flash Point 93°C (199°F)
Quantity 100g
Beilstein 1932847
Formula Weight 140.16
Percent Purity 97%
Chemical Name or Material 1-(3-Fluorophenyl)ethanol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning