Alfa Aesar™ 1-(3-Fluorophenyl)propanol, 97%

Manufacturer: Alfa Aesar™ H3234209

Quantity 10g

 View more versions of this product

Catalog No. AAH3234209

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 701-38-2
    Molecular Formula C9H11FO
    Formula Weight 154.18
    MDL Number MFCD08062380
    Chemical Name or Material 1-(3-Fluorophenyl)propanol

    Specifications

    Formula Weight 154.18
    MDL Number MFCD08062380
    Quantity 10g
    Molecular Formula C9H11FO
    CAS 701-38-2
    Chemical Name or Material 1-(3-Fluorophenyl)propanol
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit