Alfa Aesar™ 1-(3-Fluorophenyl)pyrrole, 98%

Manufacturer: Alfa Aesar™ H3334806

Quantity 5g

 View more versions of this product

Catalog No. AAH3334806

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 53871-27-5
    Molecular Formula C10H8FN
    Formula Weight 161.18
    MDL Number MFCD02665249
    Chemical Name or Material 1-(3-Fluorophenyl)pyrrole

    Specifications

    Formula Weight 161.18
    Molecular Formula C10H8FN
    MDL Number MFCD02665249
    CAS 53871-27-5
    Boiling Point 54°C (0.15mmHg)
    Quantity 5g
    Chemical Name or Material 1-(3-Fluorophenyl)pyrrole
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit