Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(3-Methoxybenzoyl)-2-(1-naphthoyl)hydrazine, 98%

Click to view available options
Quantity:
1g
5g
73941-12-5
C19H16N2O3
320.348
MFCD00021528
MMJQUHDGIXJLMO-UHFFFAOYSA-N
m-anisic acid, 1-naphthoyl hydrazide,1-m-methoxybenzoyl-2-1-naphthoyl hydrazine,hydrazine, 1-m-methoxybenzoyl-2-1-naphthoyl,1-3-methoxybenzoyl-2-1-naphthoyl hydrazine,2'-3-methoxybenzoyl-1-naphthohydrazide,1-3-methoxybenzoyl-2-1-naphthoyl-hydrazine,1-naphthalenecarboxylic acid, 2-3-methoxybenzoyl hydrazide,n'-3-methoxybenzoyl naphthalene-1-carbohydrazide,wln: l66j bvmmvr co1,n'-3-methoxybenzoyl-1-naphthohydrazide
52790
N'-(3-methoxybenzoyl)naphthalene-1-carbohydrazide
COC1=CC=CC(=C1)C(=O)NNC(=O)C2=CC=CC3=CC=CC=C32
73941-12-5
C19H16N2O3
320.348
MFCD00021528
MMJQUHDGIXJLMO-UHFFFAOYSA-N
m-anisic acid, 1-naphthoyl hydrazide,1-m-methoxybenzoyl-2-1-naphthoyl hydrazine,hydrazine, 1-m-methoxybenzoyl-2-1-naphthoyl,1-3-methoxybenzoyl-2-1-naphthoyl hydrazine,2'-3-methoxybenzoyl-1-naphthohydrazide,1-3-methoxybenzoyl-2-1-naphthoyl-hydrazine,1-naphthalenecarboxylic acid, 2-3-methoxybenzoyl hydrazide,n'-3-methoxybenzoyl naphthalene-1-carbohydrazide,wln: l66j bvmmvr co1,n'-3-methoxybenzoyl-1-naphthohydrazide
52790
N'-(3-methoxybenzoyl)naphthalene-1-carbohydrazide
COC1=CC=CC(=C1)C(=O)NNC(=O)C2=CC=CC3=CC=CC=C32
Melting Point 189°C to 191°C
Quantity 5g
Formula Weight 320.33
Percent Purity 98%
Chemical Name or Material 1-(3-Methoxybenzoyl)-2-(1-naphthoyl)hydrazine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 277-645-1

RTECSNumber : MV5370000

TSCA : No

Recommended Storage : Ambient temperatures