Alfa Aesar™ 1-(3-Methoxyphenyl)piperazine, 95%

Manufacturer: Alfa Aesar™ H5545006

Quantity 5g

 View more versions of this product

Catalog No. AAH5545006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 16015-71-7
    Molecular Formula C11H16N2O
    Formula Weight 192.26
    MDL Number MFCD00040733
    UN Number UN3267
    Chemical Name or Material 1-(3-Methoxyphenyl)piperazine

    Specifications

    Formula Weight 192.26
    MDL Number MFCD00040733
    CAS 16015-71-7
    UN Number UN3267
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.
    Molecular Formula C11H16N2O
    Boiling Point 150°C (0.5mmHg)
    Quantity 5g
    Flash Point 91°C (196°F)
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, BASIC, ORGANIC, N.O.S.
    Chemical Name or Material 1-(3-Methoxyphenyl)piperazine
    Assay 95%

    GHS H Statement
    H314-H318-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit