Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(3-Methylphenyl)piperazine, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
41186-03-2
C11H16N2
176.26
MFCD00040734
JIWHIRLNKIUYSM-UHFFFAOYSA-N
1-3-methylphenyl piperazine,1-m-tolyl piperazine,1-m-tolylpiperazine,1-3-methylphenyl-piperazine,1-m-tolyl-piperazine,piperazine, 1-3-methylphenyl,3-methylphenyl piperazine,4-m-tolylpiperazine,1-m-tolyl-piperzine,4-m-tolyl-piperazine
83111
1-(3-methylphenyl)piperazine
CC1=CC(=CC=C1)N2CCNCC2
41186-03-2
C11H16N2
176.26
MFCD00040734
JIWHIRLNKIUYSM-UHFFFAOYSA-N
1-3-methylphenyl piperazine,1-m-tolyl piperazine,1-m-tolylpiperazine,1-3-methylphenyl-piperazine,1-m-tolyl-piperazine,piperazine, 1-3-methylphenyl,3-methylphenyl piperazine,4-m-tolylpiperazine,1-m-tolyl-piperzine,4-m-tolyl-piperazine
83111
1-(3-methylphenyl)piperazine
CC1=CC(=CC=C1)N2CCNCC2
Refractive Index 1.5755 to 1.5785
Infrared Spectrum Authentic
Quantity 1g
Color Colorless to Yellow
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Formula Weight 176.26
Physical Form Viscous Liquid
Percent Purity ≥98.5%
Chemical Name or Material 1-(3-Methylphenyl)piperazine, 99%
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash face, hands and any exposed skin thoroughly after handling.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 255-251-0

TSCA : TSCA