Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,3-Propanedisulfonic acid, 70% w/v aq. soln.

Click to view available options
Quantity:
25g
100g
21668-77-9
C3H8O6S2
204.211
MFCD00191482
MGNVWUDMMXZUDI-UHFFFAOYSA-N
1,3-propanedisulfonic acid,eprodisate,1,3-propanedisulfonic acid in water,unii-6qfp76v7s7,propane-1,3-disulfonate,eprodisate inn,1,3-propanesulfonic acid,1,3-propanedisulfonicacid,1,3-propanedisulphonic acid,1,3 propanesulfonate
428573
propane-1,3-disulfonic acid
C(CS(=O)(=O)O)CS(=O)(=O)O
21668-77-9
C3H8O6S2
204.211
MFCD00191482
MGNVWUDMMXZUDI-UHFFFAOYSA-N
1,3-propanedisulfonic acid,eprodisate,1,3-propanedisulfonic acid in water,unii-6qfp76v7s7,propane-1,3-disulfonate,eprodisate inn,1,3-propanesulfonic acid,1,3-propanedisulfonicacid,1,3-propanedisulphonic acid,1,3 propanesulfonate
428573
propane-1,3-disulfonic acid
C(CS(=O)(=O)O)CS(=O)(=O)O
Quantity 25g
UN Number UN3265
Concentration or Composition (by Analyte or Components) 70% w/v aq. soln.
Formula Weight 204.21
Chemical Name or Material 1,3-Propanedisulfonic acid

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.