Alfa Aesar™ 1-[4-(4-Morpholinyl)phenyl]guanidine, 98%

Manufacturer: Alfa Aesar™ H3372614

Quantity 25g

 View more versions of this product

Catalog No. AAH3372614

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 247234-41-9
    Molecular Formula C11H16NO2
    Formula Weight 220.27
    MDL Number MFCD11986905
    Chemical Name or Material 1-[4-(4-Morpholinyl)phenyl]guanidine

    Specifications

    Formula Weight 220.27
    MDL Number MFCD11986905
    CAS 247234-41-9
    Melting Point ≈220°C (dec.)
    Molecular Formula C11H16NO2
    Quantity 25g
    Chemical Name or Material 1-[4-(4-Morpholinyl)phenyl]guanidine
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit