Alfa Aesar™ 1,4,7,10-Tetraazacyclododecane tetrahydrochloride, 98%

Manufacturer: Alfa Aesar™ B2226801

Quantity 0.25g

 View more versions of this product

Catalog No. AAB2226801

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 10045-25-7
    Molecular Formula C8H20N4·4HCl
    Formula Weight 318.12
    MDL Number MFCD00068484
    UN Number UN3077
    Synonym Cyclen tetrahydrochloride
    Chemical Name or Material 1,4,7,10-Tetraazacyclododecane tetrahydrochloride

    Specifications

    Formula Weight 318.12
    MDL Number MFCD00068484
    CAS 10045-25-7
    UN Number UN3077
    Health Hazard 3 GHS P Statement
    P280f-P273-P501a
    Wear protective clothing.
    Avoid release to the environment.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point >290°C
    Odor Odorless
    Quantity 0.25g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Molecular Formula C8H20N4·4HCl
    Synonym Cyclen tetrahydrochloride
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302
    Harmful if swallowed.
    Chemical Name or Material 1,4,7,10-Tetraazacyclododecane tetrahydrochloride
    Assay 98%

    GHS H Statement
    H302
    Harmful if swallowed.

    GHS P Statement
    P280f-P273-P501a
    Wear protective clothing.
    Avoid release to the environment.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit