Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4,7,10-Tetraazacyclododecane tetrahydrochloride, 98%

Manufacturer:  Alfa Aesar™ B2226803

Quantity 1g

 View more versions of this product

Catalog No. AAB2226803


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

10045-25-7
318.12
UN3077
1,4,7,10-Tetraazacyclododecane tetrahydrochloride
C8H20N4·4HCl
MFCD00068484
Cyclen tetrahydrochloride

Specifications

10045-25-7
318.12
UN3077
>290°C
1g
98%
C8H20N4·4HCl
MFCD00068484
Cyclen tetrahydrochloride
Odorless
1,4,7,10-Tetraazacyclododecane tetrahydrochloride

Safety and Handling

SDS & Safety Documents

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f-P273-P501a
Wear protective clothing.
Avoid release to the environment.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit