Learn More
1,4,8,11-Tetraazacyclotetradecane, 98+%, Thermo Scientific Chemicals
Description
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,4,8,11-Tetraazacyclotetradecane is used in the synthesis of molecules with electroactive cavities. It acts as a nitrogen crown ether analogue and as an antioxidant in rubber. It is also used in the preparation of plerixafor derivatives.
Solubility
Soluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications
Melting Point | 184°C to 188°C |
Quantity | 1 g |
Beilstein | 111811 |
Sensitivity | Air sensitive; Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 200.33 |
Percent Purity | ≥98% |
Chemical Name or Material | 1,4,8,11-Tetraazacyclotetradecane |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 206-039-1 |
RTECSNumber | XA5254800 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.