Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1-(4-Aminobenzyl)-1H-1,2,4-triazole, 98%

Click to view available options
Quantity:
25g
100g
119192-10-8
C9H10N4
174.207
MFCD00973306
ZGLQVRIVLWGDNA-UHFFFAOYSA-N
4-1h-1,2,4-triazol-1-ylmethyl aniline,4-1h-1,2,4-triazol-1-yl methyl aniline,1-4-aminobenzyl-1,2,4-triazole,4-1,2,4-triazol-1-ylmethyl aniline,4-1h-1,2,4-triazol-1-yl-methyl benzenamine,4-1,2,4 triazol-1-ylmethylphenylamine,benzenamine, 4-1h-1,2,4-triazol-1-ylmethyl,4-1,2,4 triazol-1-ylmethyl-phenylamine,4-1h-1,2,4-triazol-1-yl-methyl aniline,4-1h-1,2,4-triazol-1-ylmethyl benzenamine
821219
4-(1,2,4-triazol-1-ylmethyl)aniline
C1=CC(=CC=C1CN2C=NC=N2)N
119192-10-8
C9H10N4
174.207
MFCD00973306
ZGLQVRIVLWGDNA-UHFFFAOYSA-N
4-1h-1,2,4-triazol-1-ylmethyl aniline,4-1h-1,2,4-triazol-1-yl methyl aniline,1-4-aminobenzyl-1,2,4-triazole,4-1,2,4-triazol-1-ylmethyl aniline,4-1h-1,2,4-triazol-1-yl-methyl benzenamine,4-1,2,4 triazol-1-ylmethylphenylamine,benzenamine, 4-1h-1,2,4-triazol-1-ylmethyl,4-1,2,4 triazol-1-ylmethyl-phenylamine,4-1h-1,2,4-triazol-1-yl-methyl aniline,4-1h-1,2,4-triazol-1-ylmethyl benzenamine
821219
4-(1,2,4-triazol-1-ylmethyl)aniline
C1=CC(=CC=C1CN2C=NC=N2)N
Melting Point 126°C to 130°C
Color White to Yellow
Quantity 100g
UN Number UN2811
Beilstein 5934828
Formula Weight 174.21
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 1-(4-Aminobenzyl)-1H-1,2,4-triazole

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.