Learn More
Medchemexpress LLC 1,4-Benzenediacetic acid | 7325-46-4 | 96.8% | 194.19 | 25 G

Supplier: Medchemexpress LLC 25GHYW009946
1,4-Benzenediacetic acid is a chemical compound with the molecular formula C10H10O4 and a molecular weight of 194.19. It typically appears as an off-white to light brown solid and has a purity of 96.84% as determined by HPLC. This product is suitable for various laboratory applications requiring a chemical with these specifications.
- High purity of 96.84% (HPLC)
- Off-white to light brown solid appearance
- Molecular formula C10H10O4
- Molecular weight 194.19
- Recommended storage for powder: -20°C for 3 years, 4°C for 2 years
- Recommended storage in solvent: -80°C for 6 months, -20°C for 1 month
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.