Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Benzodioxan-6-amine, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
50g
250g
Packaging:
Glass bottle
22013-33-8
C8H9NO2
151.16
MFCD00006824
BZKOZYWGZKRTIB-UHFFFAOYSA-N
1,4-benzodioxan-6-amine,3,4-ethylenedioxyaniline,6-amino-1,4-benzodioxane,2,3-dihydrobenzo b 1,4 dioxin-6-amine,6-amino-1,4-benzodioxan,1,4-benzodioxan-6-ylamine,1,4-benzodioxin-6-amine, 2,3-dihydro,3,4-ethylenedioxy aniline,2,3-dihydro-benzo 1,4 dioxin-6-ylamine,2,3-dihydro-1,4-benzodioxin-6-amin
89148
2,3-dihydro-1,4-benzodioxin-6-amine
C1COC2=C(O1)C=CC(=C2)N
22013-33-8
C8H9NO2
151.16
MFCD00006824
BZKOZYWGZKRTIB-UHFFFAOYSA-N
1,4-benzodioxan-6-amine,3,4-ethylenedioxyaniline,6-amino-1,4-benzodioxane,2,3-dihydrobenzo b 1,4 dioxin-6-amine,6-amino-1,4-benzodioxan,1,4-benzodioxan-6-ylamine,1,4-benzodioxin-6-amine, 2,3-dihydro,3,4-ethylenedioxy aniline,2,3-dihydro-benzo 1,4 dioxin-6-ylamine,2,3-dihydro-1,4-benzodioxin-6-amin
89148
2,3-dihydro-1,4-benzodioxin-6-amine
C1COC2=C(O1)C=CC(=C2)N
Specific Gravity 1.23
Flash Point 120°C
Density 1.2300g/mL
Refractive Index 1.5977 to 1.5997
Quantity 250g
Color Brown to Black
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in many organic solvents
Formula Weight 151.16
Physical Form Viscous Liquid After Melting
Percent Purity 99%
Chemical Name or Material 1, 4-Benzodioxan-6-amine
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 244-718-4