Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Benzodioxan-6-yl methyl ketone, 98%, Acros Organics

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
2879-20-1
C10H10O3
178.19
MFCD00006823
HGVWMTAIIYNQSI-UHFFFAOYSA-N
6-acetyl-1,4-benzodioxane,1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone,1,4-benzodioxan-6-yl methyl ketone,1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone,1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one,6-acetyl-1,4-benzodioxan,ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl,ethanone,1-2,3-dihydro-1,4-benzodioxin-6-yl,1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethan-1-one,6-acetyl-2h,3h-benzo e 1,4-dioxin
76143
1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
CC(=O)C1=CC2=C(C=C1)OCCO2
2879-20-1
C10H10O3
178.19
MFCD00006823
HGVWMTAIIYNQSI-UHFFFAOYSA-N
6-acetyl-1,4-benzodioxane,1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethanone,1,4-benzodioxan-6-yl methyl ketone,1-2,3-dihydro-1,4-benzodioxin-6-yl ethanone,1-2,3-dihydro-1,4-benzodioxin-6-yl ethan-1-one,6-acetyl-1,4-benzodioxan,ethanone, 1-2,3-dihydro-1,4-benzodioxin-6-yl,ethanone,1-2,3-dihydro-1,4-benzodioxin-6-yl,1-2,3-dihydrobenzo b 1,4 dioxin-6-yl ethan-1-one,6-acetyl-2h,3h-benzo e 1,4-dioxin
76143
1-(2,3-dihydro-1,4-benzodioxin-6-yl)ethanone
CC(=O)C1=CC2=C(C=C1)OCCO2
Color Blue to White
Packaging Glass bottle
Melting Point 81°C to 85°C
Infrared Spectrum Authentic
Quantity 10g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 178.19
Physical Form Fine Crystalline Needles
Percent Purity 98%
Chemical Name or Material 1, 4-Benzodioxan-6-yl methyl ketone
Show More Show Less

EINECSNumber : 220-726-3