Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Benzodioxan, 99%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
493-09-4
C8H8O2
136.15
MFCD00006821
BNBQRQQYDMDJAH-UHFFFAOYSA-N
1,4-benzodioxane,1,4-benzodioxan,2,3-dihydrobenzo b 1,4 dioxine,1,4-benzodioxin, 2,3-dihydro,pyrocatechol ethylene ether,1,2-ethylenedioxy benzene,benzodioxan,ethylene o-phenylene dioxide,1,2-ethylenedioxybenzene,1, 4-benzdioxan
10301
2,3-dihydro-1,4-benzodioxine
C1COC2=CC=CC=C2O1
493-09-4
C8H8O2
136.15
MFCD00006821
BNBQRQQYDMDJAH-UHFFFAOYSA-N
1,4-benzodioxane,1,4-benzodioxan,2,3-dihydrobenzo b 1,4 dioxine,1,4-benzodioxin, 2,3-dihydro,pyrocatechol ethylene ether,1,2-ethylenedioxy benzene,benzodioxan,ethylene o-phenylene dioxide,1,2-ethylenedioxybenzene,1, 4-benzdioxan
10301
2,3-dihydro-1,4-benzodioxine
C1COC2=CC=CC=C2O1
Color Colorless to Yellow
Specific Gravity 1.14
Quantity 100mL
Flash Point 87°C
Boiling Point 248°C
Refractive Index 1.5485 to 1.5505
Infrared Spectrum Authentic
Density 1.1400g/mL
Packaging Glass bottle
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Beilstein 19, 22
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in all typical organic solvents
Formula Weight 136.15
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material 1, 4-Benzodioxan
Show More Show Less

EINECSNumber : 207-775-6