Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 1,4-Bis(2-hydroxyethyl)piperazine, 98%

Manufacturer:  Alfa Aesar™ H5506306

Quantity 5g

 View more versions of this product

Catalog No. AAH5506306


  Check Availability
Add to cart

Specifications

Description

Applications
It is applied for desulfurization, decarburization extraction.

Solubility
Soluble in water.

Notes
Store at 4°C. Keep away from oxidizing agents.
Description & Specifications

Chemical Identifiers

122-96-3
174.24
1,4-Piperazinediethanol
C8H18N2O2
MFCD00006157
1,4-Bis(2-hydroxyethyl)piperazine

Specifications

122-96-3
174.24
1,4-Piperazinediethanol
215° to 220°C (50mmHg)
1,4-Bis(2-hydroxyethyl)piperazine
C8H18N2O2
MFCD00006157
133° to 136°C
5g
99%

Safety and Handling

SDS & Safety Documents

RTECSNumber : TL3675000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit