Alfa Aesar™ 1,4-Bis(2-phenylethyl)benzene, 97%

Manufacturer: Alfa Aesar™ H3033803

Quantity 1g

 View more versions of this product

Catalog No. AAH3033803

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 1985-58-6
    Molecular Formula C22H22
    Formula Weight 286.42
    MDL Number MFCD12407076
    Synonym 1,4-Diphenethylbenzene; 4-Phenethylbibenzyl
    Chemical Name or Material 1,4-Bis(2-phenylethyl)benzene

    Specifications

    Formula Weight 286.42
    Molecular Formula C22H22
    MDL Number MFCD12407076
    CAS 1985-58-6
    Synonym 1,4-Diphenethylbenzene; 4-Phenethylbibenzyl
    Melting Point 88° to 90°C
    Quantity 1g
    Odor Odorless
    Chemical Name or Material 1,4-Bis(2-phenylethyl)benzene
    Assay 97%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit