Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis(trichloromethyl)benzene, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
68-36-0
C8H4Cl6
312.83
MFCD00000791
OTEKOJQFKOIXMU-UHFFFAOYSA-N
1,4-bis trichloromethyl benzene,bitriben,chloksil,chloxil,chloxyl,hexichol,khloksil,khloxil,cloxil,hetol
6233
1,4-bis(trichloromethyl)benzene
C1=CC(=CC=C1C(Cl)(Cl)Cl)C(Cl)(Cl)Cl
68-36-0
C8H4Cl6
312.83
MFCD00000791
OTEKOJQFKOIXMU-UHFFFAOYSA-N
1,4-bis trichloromethyl benzene,bitriben,chloksil,chloxil,chloxyl,hexichol,khloksil,khloxil,cloxil,hetol
6233
1,4-bis(trichloromethyl)benzene
C1=CC(=CC=C1C(Cl)(Cl)Cl)C(Cl)(Cl)Cl
Viscosity 2.07 mPa.s (120°C)
Boiling Point 312°C
Melting Point 106°C to 110°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
Quantity 1g
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 98.5% min. (GC)
Linear Formula C6H4(CCl3)2
Beilstein 05, 385
Merck Index 15, 1304
Solubility Information Solubility in water: insoluble
Formula Weight 312.83
Physical Form Crystalline Powder and Chunks
Percent Purity 99%
Chemical Name or Material 1, 4-Bis(trichloromethyl)benzene, 99%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 200-686-3

RTECSNumber : ZE4655000

TSCA : TSCA