Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,4-Bis[(trimethylsilyl)ethynyl]benzene 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
17938-13-5
C16H22Si2
270.522
MFCD00135433
CMTMWEXUJQSPCA-UHFFFAOYSA-N
1,4-bis trimethylsilyl ethynyl benzene,1,4-bis trimethylsilylethynyl benzene,1,4-bis 2-trimethylsilyl ethynyl benzene,trimethyl-2-4-2-trimethylsilylethynyl phenyl ethynyl silane,trimethyl 2-4-2-trimethylsilyl ethynyl phenyl ethynyl silane,acmc-20ap2w,trimethyl 4-trimethylsilyl ethynyl phenyl ethynyl silane,benzene,1,4-bis 2-trimethylsilyl ethynyl
522492
trimethyl-[2-[4-(2-trimethylsilylethynyl)phenyl]ethynyl]silane
C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C
17938-13-5
C16H22Si2
270.522
MFCD00135433
CMTMWEXUJQSPCA-UHFFFAOYSA-N
1,4-bis trimethylsilyl ethynyl benzene,1,4-bis trimethylsilylethynyl benzene,1,4-bis 2-trimethylsilyl ethynyl benzene,trimethyl-2-4-2-trimethylsilylethynyl phenyl ethynyl silane,trimethyl 2-4-2-trimethylsilyl ethynyl phenyl ethynyl silane,acmc-20ap2w,trimethyl 4-trimethylsilyl ethynyl phenyl ethynyl silane,benzene,1,4-bis 2-trimethylsilyl ethynyl
522492
trimethyl-[2-[4-(2-trimethylsilylethynyl)phenyl]ethynyl]silane
C[Si](C)(C)C#CC1=CC=C(C=C1)C#C[Si](C)(C)C
Packaging Glass bottle
Melting Point 119°C to 123°C
Quantity 5g
Infrared Spectrum Authentic
Color White
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula C6H4[CCSi(CH3)3]2
Formula Weight 270.52
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 1, 4-Bis[(trimethylsilyl)ethynyl]benzene
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.